Stampa…
Anti-YWHAE Rabbit Polyclonal Antibody
Fornitore: Biorbyt
BIRBORB48976-200EA 0 EUR
BIRBORB48976-200 BIRBORB48976-100
Anti-YWHAE Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: YWHAE (tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, epsilon)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat