Stampa…

Anti-P4HB Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB48924-100EA 0 EUR
BIRBORB48924-100 BIRBORB48924-200
Anti-P4HB Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: P4HB (prolyl 4-hydroxylase, beta polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR