Stampa…

Anti-LGALS3 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB48890-200EA 0 EUR
BIRBORB48890-200
Anti-LGALS3 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: LGALS3 (lectin, galactoside-binding, soluble, 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR