Stampa…

Anti-BCL2L2 Rabbit polyclonal antibody

Fornitore: Biorbyt
BIRBORB48800-200EA 0 EUR
BIRBORB48800-200
Anti-BCL2L2 Rabbit polyclonal antibody
Anticorpi
Type: Primary
Antigen: BCL2L2 (BCL2-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR