Stampa…

Anti-MSLN Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB48779-200EA 0 EUR
BIRBORB48779-200
Anti-MSLN Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: MSLN (mesothelin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR