Stampa…

Anti-MYOSTP1 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB48771-200EA 0 EUR
BIRBORB48771-200
Anti-MYOSTP1 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: MYOSTP1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR