Stampa…

Anti-MAPT Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB48688-200EA 0 EUR
BIRBORB48688-200
Anti-MAPT Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG (Mouse)
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR