Stampa…

Anti-IL17F Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB48666-200EA 0 EUR
BIRBORB48666-200
Anti-IL17F Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: IL17F (interleukin 17F)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR