Stampa…

Anti-TG Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB48647-200EA 0 EUR
BIRBORB48647-200
Anti-TG Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: TG (thyroglobulin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR