Stampa…

Anti-MAP2K7 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB48447-100EA 0 EUR
BIRBORB48447-100
Anti-MAP2K7 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: MAP2K7 (mitogen-activated protein kinase kinase 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR