Stampa…

Anti-GGT1 Rabbit polyclonal antibody

Fornitore: Biorbyt
BIRBORB48385-100EA 0 EUR
BIRBORB48385-100
Anti-GGT1 Rabbit polyclonal antibody
Anticorpi
Type: Primary
Antigen: GGT1 (gamma-glutamyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR