Stampa…

Anti-G6PD Rabbit polyclonal antibody

Fornitore: Biorbyt
BIRBORB48382-100EA 0 EUR
BIRBORB48382-100
Anti-G6PD Rabbit polyclonal antibody
Anticorpi
Type: Primary
Antigen: G6PD (glucose-6-phosphate dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR