Stampa…

Anti-CYP11A1 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB48342-100EA 0 EUR
BIRBORB48342-100
Anti-CYP11A1 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: CYP11A1 (cytochrome P450, family 11, subfamily A, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR