Stampa…

Anti-CD3G Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB48108-100EA 0 EUR
BIRBORB48108-100
Anti-CD3G Mouse Monoclonal Antibody
Anticorpi
Western Blot: 1:500-1:1000; Immunofluorescence: 1:100-1:500

Type: Primary
Antigen: CD3G (CD3g molecule, gamma (CD3-TCR complex))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG (Mouse)
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR