Stampa…

Anti-GBP1 Rabbit polyclonal antibody

Fornitore: Biorbyt
BIRBORB48017-100EA 0 EUR
BIRBORB48017-100
Anti-GBP1 Rabbit polyclonal antibody
Anticorpi
Type: Primary
Antigen: GBP1 (guanylate binding protein 1, interferon-inducible)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR