Stampa…

Anti-IL4 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB47996-100EA 0 EUR
BIRBORB47996-100
Anti-IL4 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: IL4 (interleukin 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR