Stampa…

Anti-CCR10 Rabbit polyclonal antibody

Fornitore: Biorbyt
BIRBORB47958-100EA 0 EUR
BIRBORB47958-100
Anti-CCR10 Rabbit polyclonal antibody
Anticorpi
Type: Primary
Antigen: CCR10 (chemokine (C-C motif) receptor 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR