Stampa…

Anti-EGFRvIII Mouse monoclonal antibody

Fornitore: Biorbyt
BIRBORB47907-100EA 0 EUR
BIRBORB47907-100
Anti-EGFRvIII Mouse monoclonal antibody
Anticorpi
Immunochemistry: 1:250-500; Immunofluorescence: 1:500

Type: Primary
Antigen: EGFRvIII
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR