Stampa…

Anti-IgM Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Fornitore: Biorbyt
BIRBORB44861-0.1EA 0 EUR
BIRBORB44861-0.1
Anti-IgM Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpi
Type: Secondary
Antigen: IgM (Human) (human IgM)
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR