Stampa…

Anti-MHC Class II Rat Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB44830-0.1EA 0 EUR
BIRBORB44830-0.1
Anti-MHC Class II Rat Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: MHC Class II
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR