Stampa…

Anti-ITGB2 Rat Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB44827-0.1EA 0 EUR
BIRBORB44827-0.1
Anti-ITGB2 Rat Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: ITGB2 (integrin, beta 2 (complement component 3 receptor 3 and 4 subunit))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR