Stampa…
Anti-FOXP3 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Fornitore: Biorbyt
BIRBORB44650-0.1EA 0 EUR
BIRBORB44650-0.1
Anti-FOXP3 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Anticorpi
Type: Primary
Antigen: FOXP3 (forkhead box P3)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici