Stampa…

Anti-CD3 Hamster Monoclonal Antibody (FITC (Fluorescein))

Fornitore: Biorbyt
BIRBORB44594-0.1EA 0 EUR
BIRBORB44594-0.1
Anti-CD3 Hamster Monoclonal Antibody (FITC (Fluorescein))
Anticorpi
Type: Primary
Antigen: CD3
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR