Stampa…

Anti-TNFRSF10C Mouse Monoclonal Antibody (PE (Phycoerythrin))

Fornitore: Biorbyt
BIRBORB44522-0.1EA 0 EUR
BIRBORB44522-0.1
Anti-TNFRSF10C Mouse Monoclonal Antibody (PE (Phycoerythrin))
Anticorpi
Type: Primary
Antigen: TNFRSF10C (tumor necrosis factor receptor superfamily, member 10c, decoy without an intracellular domain)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR