Stampa…

Anti-ITGAL Mouse Monoclonal Antibody (FITC (Fluorescein))

Fornitore: Biorbyt
BIRBORB44221-100EA 0 EUR
BIRBORB44221-100
Anti-ITGAL Mouse Monoclonal Antibody (FITC (Fluorescein))
Anticorpi
Type: Primary
Antigen: ITGAL (integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide))
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR