Stampa…

Anti-TFRC Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB44174-0.1EA 0 EUR
BIRBORB44174-0.1
Anti-TFRC Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: TFRC (transferrin receptor)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR