Stampa…

Anti-ITGAM Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB44136-0.1EA 0 EUR
BIRBORB44136-0.1
Anti-ITGAM Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: ITGAM (integrin, alpha M (complement component 3 receptor 3 subunit))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR