Stampa…

Anti-ITGB2 Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB44079-0.1EA 0 EUR
BIRBORB44079-0.1
Anti-ITGB2 Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: ITGB2 (integrin, beta 2 (complement component 3 receptor 3 and 4 subunit))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR