Stampa…

Anti-HLA-DR Mouse Monoclonal Antibody (FITC (Fluorescein))

Fornitore: Biorbyt
BIRBORB44043-100EA 0 EUR
BIRBORB44043-100
Anti-HLA-DR Mouse Monoclonal Antibody (FITC (Fluorescein))
Anticorpi
Type: Primary
Antigen: HLA-DR
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR