Stampa…

Anti-MS4A1 Mouse Monoclonal Antibody (FITC (Fluorescein))

Fornitore: Biorbyt
BIRBORB43939-100EA 0 EUR
BIRBORB43939-100
Anti-MS4A1 Mouse Monoclonal Antibody (FITC (Fluorescein))
Anticorpi
Type: Primary
Antigen: MS4A1 (membrane-spanning 4-domains, subfamily A, member 1)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR