Stampa…

Anti-CD22 Mouse Monoclonal Antibody (FITC (Fluorescein))

Fornitore: Biorbyt
BIRBORB43883-100EA 0 EUR
BIRBORB43883-100
Anti-CD22 Mouse Monoclonal Antibody (FITC (Fluorescein))
Anticorpi
Type: Primary
Antigen: CD22 (CD22 molecule)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR