Stampa…

Anti-Cytokeratin 7/17 Mouse monoclonal antibody

Fornitore: Biorbyt
BIRBORB43713-0.1EA 0 EUR
BIRBORB43713-0.1
Anti-Cytokeratin 7/17 Mouse monoclonal antibody
Anticorpi
Type: Primary
Antigen: Cytokeratin 7/17
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR