Stampa…

Anti-Clathrin (H chain) Mouse monoclonal antibody

Fornitore: Biorbyt
BIRBORB43680-0.1EA 0 EUR
BIRBORB43680-0.1
Anti-Clathrin (H chain) Mouse monoclonal antibody
Anticorpi
Type: Primary
Antigen: Clathrin (H chain)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR