Stampa…

Anti-PLSCR1 Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB43576-0.1EA 0 EUR
BIRBORB43576-0.1
Anti-PLSCR1 Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: PLSCR1 (phospholipid scramblase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR