Stampa…

Anti-IGFBP3 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB43510-100EA 0 EUR
BIRBORB43510-100
Anti-IGFBP3 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: IGFBP3 (insulin-like growth factor binding protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR