Stampa…

Anti-MAPT Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB43355-100EA 0 EUR
BIRBORB43355-100
Anti-MAPT Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR