Stampa…

Anti-SUFU Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB43350-100EA 0 EUR
BIRBORB43350-100
Anti-SUFU Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: SUFU (suppressor of fused homolog (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR