Stampa…

Anti-His tag Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB43332-100EA 0 EUR
BIRBORB43332-100
Anti-His tag Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: His tag (His tag)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR