Stampa…

Anti-ZWILCH Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB43317-100EA 0 EUR
BIRBORB43317-100
Anti-ZWILCH Rabbit Polyclonal Antibody
Anticorpi
Western Blot: 1:1000

Type: Primary
Antigen: ZWILCH (zwilch kinetochore protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR