Stampa…

Anti-LCN1 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB42821-200EA 0 EUR
BIRBORB42821-200
Anti-LCN1 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: LCN1 (lipocalin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR