Stampa…

Anti-CD74 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB41626-100EA 0 EUR
BIRBORB41626-100
Anti-CD74 Rabbit Polyclonal Antibody
Anticorpi
Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:500

Type: Primary
Antigen: CD74 (CD74 molecule, major histocompatibility complex, class II invariant chain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR