Stampa…

Anti-DHH Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB41565-100EA 0 EUR
BIRBORB41565-100
Anti-DHH Rabbit Polyclonal Antibody
Anticorpi
Western Blot: 1:500-1:3000

Type: Primary
Antigen: DHH (desert hedgehog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR