Stampa…

Anti-PIK3R4 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB41353-100EA 0 EUR
BIRBORB41353-100
Anti-PIK3R4 Rabbit Polyclonal Antibody
Anticorpi
Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: PIK3R4 (phosphoinositide-3-kinase, regulatory subunit 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR