Stampa…
Anti-Interferon alpha 8 Rabbit Polyclonal Antibody
Fornitore: Biorbyt
BIRBORB41174-100EA 0 EUR
BIRBORB41174-100
Anti-Interferon alpha 8 Rabbit Polyclonal Antibody
Anticorpi
Western Blot: 1:500-1:3000
Type: Primary
Antigen: Interferon alpha 8
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human