Stampa…

Anti-COL1A2 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB40955-100EA 0 EUR
BIRBORB40955-100
Anti-COL1A2 Rabbit Polyclonal Antibody
Anticorpi
Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: COL1A2 (collagen, type I, alpha 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR