Stampa…

Anti-FABP5 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB27768-100EA 0 EUR
BIRBORB27768-100
Anti-FABP5 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: FABP5 (fatty acid binding protein 5 (psoriasis-associated))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR