Stampa…

Anti-CXCL16 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB27652-100EA 0 EUR
BIRBORB27652-100
Anti-CXCL16 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: CXCL16 (chemokine (C-X-C motif) ligand 16)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR