Stampa…

Anti-MGMT None Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB27618-100EA 0 EUR
BIRBORB27618-100
Anti-MGMT None Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: MGMT (O-6-methylguanine-DNA methyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: IgG (Rabbit)
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR