Stampa…

Anti-LCN2 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
BIRBORB27593-100EA 0 EUR
BIRBORB27593-100
Anti-LCN2 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: LCN2 (lipocalin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR