Stampa…

Anti-Pentosidine Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB27502-50EA 0 EUR
BIRBORB27502-50
Anti-Pentosidine Mouse Monoclonal Antibody
Anticorpi
ELISA: 0.1-0.5 ?g/ml; Immunochemistry: 2 ?g/ml

Type: Primary
Antigen: Pentosidine
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR